Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-pentanol, 96%

Chemical Identifiers

Chemical Name or Material 5-Amino-1-pentanol
CAS 2508-29-4
Molecular Formula C5H13NO
Linear Formula H2N(CH2)5OH
Formula Weight 103.16g/mol
MDL Number MFCD00008237
UN Number UN3259
Beilstein 1732302
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0197914

 
alfa aesar™
L01979-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 34° to 37°C
Boiling Point 222°C
Quantity 25g
Flash Point >100°C (212°F)
Chemical Name or Material 5-Amino-1-pentanol
CAS 2508-29-4
Assay 96%
Molecular Formula C5H13NO
Linear Formula H2N(CH2)5OH
Formula Weight 103.16g/mol
MDL Number MFCD00008237
UN Number UN3259
Beilstein 1732302
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.