missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-methyl-3-phenyl-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ A1575414

 View more versions of this product

Catalog No. AAA1575414



Specifications

Specifications

5-Amino-1-methyl-3-phenyl-1H-pyrazole
124°C to 130°C
10199-50-5
MFCD00067874
1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
CN1C(=CC(=N1)C2=CC=CC=C2)N
173.219
173.22
Odorless
25g
C10H11N3
743807
KCYRMURRLLYLPU-UHFFFAOYSA-N
2-methyl-5-phenylpyrazol-3-amine
517779
97%

Chemical Identifiers

10199-50-5
173.219
KCYRMURRLLYLPU-UHFFFAOYSA-N
517779
CN1C(=CC(=N1)C2=CC=CC=C2)N
C10H11N3
MFCD00067874
1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
2-methyl-5-phenylpyrazol-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents