missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde, 97%, Maybridge

Manufacturer:  Thermo Scientific MO07848DA

 View more versions of this product

Catalog No. MO07848DA



Specifications

Specifications

5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
1g
97.0
C5H7N3O
5-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 5-amino-1-methylpyrazole-4-carboxaldehyde, 5-amino-1-methyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 5-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 5-amino-1-methyl-9ci
CN1C(=C(C=N1)C=O)N
125.131
125.13
Amber Glass Bottle
62564-90-3
97%
MFCD08690324
NDGIYFSWJGZTRC-UHFFFAOYSA-N
5-amino-1-methylpyrazole-4-carbaldehyde
12332515
97%

Chemical Identifiers

62564-90-3
125.131
NDGIYFSWJGZTRC-UHFFFAOYSA-N
12332515
CN1C(=C(C=N1)C=O)N
C5H7N3O
MFCD08690324
5-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 5-amino-1-methylpyrazole-4-carboxaldehyde, 5-amino-1-methyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 5-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 5-amino-1-methyl-9ci
5-amino-1-methylpyrazole-4-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents