missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde, 97%, Maybridge

$597.03 - $597.03

Chemical Identifiers

CAS 62564-90-3
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C5H7N3O
Formula Weight 125.13
MDL Number MFCD08690324
Chemical Name or Material 5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO07848DA SDS Fisher Scientific Company
MO07848DA
Amber Glass Bottle 1g Each for $597.03
Specifications Description & Specifications

Chemical Identifiers

62564-90-3
97%
125.13
5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
97.0
C5H7N3O
MFCD08690324
97%

Specifications

125.13
97.0
C5H7N3O
MFCD08690324
5-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
1g
Amber Glass Bottle
62564-90-3
97%
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.