Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-indanone, 95%

Chemical Identifiers

Chemical Name or Material 5-Amino-1-indanone
CAS 3470-54-0
Formula Weight 147.18
MDL Number MFCD00099466
UN Number UN2811
Beilstein 1636158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6642503

  SDS
alfa aesar™
H6642503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66425MD

  SDS
alfa aesar™
H66425MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 184° to 186°C
Formula Weight 147.18
Chemical Name or Material 5-Amino-1-indanone
Assay 95%
CAS 3470-54-0
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Beilstein 1636158
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
UN Number UN2811
Quantity 1g
MDL Number MFCD00099466
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.