missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1-ethylpyrazole, 99%, Acros Organics

Chemical Identifiers

Assay Percent Range 99%
Molecular Formula C5H9N3
Formula Weight 111.15
Chemical Name or Material 5-Amino-1-ethylpyrazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC437620050 Thermo Fisher Scientific
AC437620050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437620010 SDS Thermo Fisher Scientific
AC437620010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

99%
C5H9N3
111.15
5-Amino-1-ethylpyrazole

Specifications

3528-58-3
99%
111.15
5g
C5H9N3
5-Amino-1-ethylpyrazole