missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Amino-1-ethylpyrazole, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437620050
Specifications
5-Amino-1-ethylpyrazole | |
106°C to 108°C (4.0 Torr) | |
5g | |
98.5% min. (GC) | |
1-ethyl-1h-pyrazol-5-amine, 1-ethyl-5-aminopyrazol, 1-ethyl-5-aminopyrazole, 1-ethylpyrazole-5-ylamine, 1h-pyrazol-5-amine, 1-ethyl, 2-ethyl-2h-pyrazol-3-ylamine, 2-ethyl-3-aminopyrazole, 5-amino-1-ethyl-1h-pyrazole, 5-amino-1-ethyl-pyrazole, 5-amino-1-ethylpyrazole | |
CCN1C(=CC=N1)N | |
111.15 | |
111.15 |
>113°C | |
54°C to 57°C | |
3528-58-3 | |
C5H9N3 | |
FSJOLBAFVKSQQJ-UHFFFAOYSA-N | |
2-ethylpyrazol-3-amine | |
337310 | |
99% |
Chemical Identifiers
3528-58-3 | |
111.15 | |
1-ethyl-1h-pyrazol-5-amine, 1-ethyl-5-aminopyrazol, 1-ethyl-5-aminopyrazole, 1-ethylpyrazole-5-ylamine, 1h-pyrazol-5-amine, 1-ethyl, 2-ethyl-2h-pyrazol-3-ylamine, 2-ethyl-3-aminopyrazole, 5-amino-1-ethyl-1h-pyrazole, 5-amino-1-ethyl-pyrazole, 5-amino-1-ethylpyrazole | |
2-ethylpyrazol-3-amine |
C5H9N3 | |
FSJOLBAFVKSQQJ-UHFFFAOYSA-N | |
337310 | |
CCN1C(=CC=N1)N |
Safety and Handling
GHS Signal Word: Warning