Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole, 97%

$220.12 - $220.12

Chemical Identifiers

Chemical Name or Material 5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole
CAS 40401-39-6
Molecular Formula C10H10ClN3
Formula Weight 207.66
MDL Number MFCD03478180
Synonym 2-(4-Chlorophenyl)-5-methyl-2H-pyrazol-3-ylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5108503

 
alfa aesar™
H5108503
1g Each for $220.12
Description & Specifications

Specifications

Formula Weight 207.66
Chemical Name or Material 5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole
TSCA TSCA
Quantity 1g
Assay 97%
Molecular Formula C10H10ClN3
MDL Number MFCD03478180
CAS 40401-39-6
Synonym 2-(4-Chlorophenyl)-5-methyl-2H-pyrazol-3-ylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.