missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 155972500

 View more versions of this product

Catalog No. AC155972500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Amino-1, 3, 4-thiadiazole-2-thiol
1% max. (110°C)
Cream to Yellow or White
250g
2349-67-9
100.0
C2H3N3S2
27, 674
Solubility in water: insoluble. Other solubilities: soluble in nh4oh,soluble in dmf
C1(=NNC(=S)S1)N
133.2
133.2
98%
Authentic
Glass bottle
231°C to 236°C
98%
97.5
97.5% min. (HPLC)
MFCD00003108
1,3,4-thiadiazole-2 3h-thione, 5-amino
GDGIVSREGUOIJZ-UHFFFAOYSA-N
5-amino-3H-1,3,4-thiadiazole-2-thione
2723847
Powder

Chemical Identifiers

2349-67-9
133.2
GDGIVSREGUOIJZ-UHFFFAOYSA-N
2723847
C1(=NNC(=S)S1)N
C2H3N3S2
MFCD00003108
1,3,4-thiadiazole-2 3h-thione, 5-amino
5-amino-3H-1,3,4-thiadiazole-2-thione
Safety and Handling

Safety and Handling

EINECSNumber : 219-078-4

RTECSNumber : XI4550000

TSCA : TSCA

SDS Documents