missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 155970250

 View more versions of this product

Catalog No. AC155970250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Amino-1, 3, 4-thiadiazole-2-thiol
2349-67-9
100.0
Cream to Yellow or White
Authentic
Glass bottle
MFCD00003108
27, 674
Solubility in water: insoluble. Other solubilities: soluble in nh4oh,soluble in dmf
C1(=NNC(=S)S1)N
133.2
133.2
Powder
98%
97.5
231°C to 236°C
1% max. (110°C)
97.5% min. (HPLC)
C2H3N3S2
25 g
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
GDGIVSREGUOIJZ-UHFFFAOYSA-N
5-amino-3H-1,3,4-thiadiazole-2-thione
2723847
98%

Chemical Identifiers

2349-67-9
133.2
GDGIVSREGUOIJZ-UHFFFAOYSA-N
2723847
C1(=NNC(=S)S1)N
C2H3N3S2
MFCD00003108
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
5-amino-3H-1,3,4-thiadiazole-2-thione
Safety and Handling

Safety and Handling

EINECSNumber : 219-078-4

RTECSNumber : XI4550000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only