Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Amino-1-(2-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%

$72.64 - $235.05

Chemical Identifiers

Chemical Name or Material 5-Amino-1-(2-chlorophenyl)-1H-pyrazole-4-carbonitrile
CAS 64096-89-5
Molecular Formula C10H7ClN4
Formula Weight 218.65
MDL Number MFCD00128297
Synonym 5-Amino-1-(2-chlorophenyl)-4-cyano-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705503

  SDS
alfa aesar™
H2705503
1g Each for $72.64

AAH2705506

  SDS
alfa aesar™
H2705506
5g Each for $235.05
Description & Specifications

Specifications

Formula Weight 218.65
Assay 97%
MDL Number MFCD00128297
Quantity 1g
Melting Point 136° to 140°C
Molecular Formula C10H7ClN4
Health Hazard 1 Danger
Synonym 5-Amino-1-(2-chlorophenyl)-4-cyano-1H-pyrazole
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 5-Amino-1-(2-chlorophenyl)-1H-pyrazole-4-carbonitrile
CAS 64096-89-5
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.