Please login to your online account to display your discounted pricing

5-Amino-1,2,4-thiadiazole, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Amino-1, 2, 4-thiadiazole
Assay Percent Range 97%
Molecular Formula C2H3N3S
Formula Weight 101.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC449870050

  SDS
thermo fisher scientific
AC449870050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC449870010

  SDS
thermo fisher scientific
AC449870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.13
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C2H3N3S
Chemical Name or Material 5-Amino-1, 2, 4-thiadiazole
CAS 7552-07-0