missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetylvaleric acid, 96%

Manufacturer:  Alfa Aesar™ A1481006

 View more versions of this product

Catalog No. AAA1481006



Description

Description

Applications
5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
Specifications

Specifications

5-Acetylvaleric acid
34°C to 36°C
3128-07-2
MFCD00051479
1362931
Soluble in water
CC(=O)CCCCC(=O)O
144.17
144.17
138°C to 140°C (1.5mmHg)
5g
C7H12O3
UN3261
5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
IZOQMUVIDMLRDC-UHFFFAOYSA-N
6-oxoheptanoic acid
98810
96%

Chemical Identifiers

3128-07-2
144.17
IZOQMUVIDMLRDC-UHFFFAOYSA-N
98810
CC(=O)CCCCC(=O)O
C7H12O3
MFCD00051479
5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
6-oxoheptanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents