Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetyluracil, 98%

$1,463.07 - $1,463.07

Chemical Identifiers

Chemical Name or Material 5-Acetyluracil
CAS 6214-65-9
Molecular Formula C6H6N2O3
Formula Weight 154.11
MDL Number MFCD00151968
Synonym 5-Acetyl-1H-pyrimidine-2,4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4437814

  SDS
alfa aesar™
4437814
25g Each for $1,463.07
Description & Specifications

Specifications

Formula Weight 154.11
Quantity 25g
Assay 98%
Chemical Name or Material 5-Acetyluracil
Physical Form Powder
Synonym 5-Acetyl-1H-pyrimidine-2,4-dione
CAS 6214-65-9
Molecular Formula C6H6N2O3
MDL Number MFCD00151968