missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetyluracil, 98%

Manufacturer:  Alfa Aesar™ 4437814

 View more versions of this product

Catalog No. AA4437814



Specifications

Specifications

5-Acetyluracil
6214-65-9
MFCD00151968
YNYDWEIQSDFDLK-UHFFFAOYSA-N
5-acetyl-1H-pyrimidine-2,4-dione
234581
Powder
25g
C6H6N2O3
1-2,4-dihydroxypyrimidin-5-yl ethanone
CC(=O)C1=CNC(=O)NC1=O
154.125
154.11
98%

Chemical Identifiers

6214-65-9
154.125
YNYDWEIQSDFDLK-UHFFFAOYSA-N
234581
CC(=O)C1=CNC(=O)NC1=O
C6H6N2O3
MFCD00151968
1-2,4-dihydroxypyrimidin-5-yl ethanone
5-acetyl-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents