Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetylthiophene-2-carbonitrile, 97%

Chemical Identifiers

Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
CAS 88653-55-8
Molecular Formula C7H5NOS
Formula Weight 151.19
MDL Number MFCD00052314
UN Number UN3439
Beilstein 1306961
Synonym 2-Acetyl-5-cyanothiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1276502

  SDS
alfa aesar™
A1276502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.19
Boiling Point 95° to 97°C (0.5mmHg)
Assay 97%
Health Hazard 1 Danger
Synonym 2-Acetyl-5-cyanothiophene
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
MDL Number MFCD00052314
Molecular Formula C7H5NOS
Beilstein 1306961
Quantity 0.5g
Melting Point 76° to 80°C
CAS 88653-55-8
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.