Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetylsalicylamide, 98%

$33.00 - $189.85

Chemical Identifiers

Chemical Name or Material 5-Acetylsalicylamide
CAS 40187-51-7
Molecular Formula C9H9NO3
Formula Weight 179.18
MDL Number MFCD00049222
Synonym 5-Acetyl-2-hydroxybenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1830306

  SDS
alfa aesar™
A1830306
5g Each for $33.00

AAA1830322

  SDS
alfa aesar™
A1830322
100g Each for $189.85
Description & Specifications

Specifications

Formula Weight 179.18
CAS 40187-51-7
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number CU8702280
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Molecular Formula C9H9NO3
Synonym 5-Acetyl-2-hydroxybenzamide
Chemical Name or Material 5-Acetylsalicylamide
MDL Number MFCD00049222
Melting Point 220° to 222°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.