Please login to your online account to display your discounted pricing

5-Acetyl-2-thiophenecarbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Acetyl-2-thiophenecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H6O2S
Formula Weight 154.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO01240DA

  SDS
fisher scientific company
MO01240DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.19
Quantity 1g
Assay 97%
Chemical Name or Material 5-Acetyl-2-thiophenecarbaldehyde
Assay Percent Range 97%
CAS 4565-29-1
Molecular Formula C7H6O2S