Please login to your online account to display your discounted pricing

5-Acetyl-2-thiophenecarbaldehyde, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  MO01240DA

 View more versions of this product

Catalog No. MO01240DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.19
Quantity 1g
Assay 97%
Chemical Name or Material 5-Acetyl-2-thiophenecarbaldehyde
Assay Percent Range 97%
CAS 4565-29-1
Molecular Formula C7H6O2S

Specifications

Chemical Name or Material 5-Acetyl-2-thiophenecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H6O2S
Formula Weight 154.19