Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetyl-2-methylpyridine, 97%

$138.54 - $346.34

Chemical Identifiers

Chemical Name or Material 5-Acetyl-2-methylpyridine
CAS 36357-38-7
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00128113
Synonym 5-Acetyl-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2701403

 
alfa aesar™
H2701403
1g Each for $346.34

AAH27014MD

 
alfa aesar™
H27014MD
250mg Each for $138.54
Description & Specifications

Specifications

Formula Weight 135.17
Chemical Name or Material 5-Acetyl-2-methylpyridine
TSCA TSCA
Assay 97%
Molecular Formula C8H9NO
MDL Number MFCD00128113
CAS 36357-38-7
Synonym 5-Acetyl-2-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
5-Acetyl-2-methylpyridine is used in the preparation of pyridine-2,5-dicarboxylic acid by using potassium permanganate as an oxidant. Further, it serves as a ligand for Suzuki coupling.

Solubility
Partly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.