Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetyl-2,3-dihydrobenzo[b]furan, 97%

$86.95 - $86.95

Chemical Identifiers

Chemical Name or Material 5-Acetyl-2,3-dihydrobenzo[b]furan
CAS 90843-31-5
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00068033
Beilstein 131198
Synonym 5-Acetylcoumaran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2026502

  SDS
alfa aesar™
B2026502
0.5g Each for $86.95

AAB2026509

  SDS
alfa aesar™
B2026509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Acetyl-2,3-dihydrobenzo[b]furan
CAS 90843-31-5
Assay 97%
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00068033
Beilstein 131198
Synonym 5-Acetylcoumaran
Boiling Point 120° to 121°C (15mmHg)
Melting Point 56° to 59°C
Quantity 0.5g