missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Acetyl-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific B2026502

 View more versions of this product

Catalog No. AAB2026502



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Acetyl-2,3-dihydrobenzo[b]furan
56°C to 59°C
C10H10O2
131198
1-2,3-dihydro-1-benzofuran-5-yl ethan-1-one, 1-2,3-dihydro-1-benzofuran-5-yl ethanone, 1-2,3-dihydro-5-benzofuranyl ethanone, 1-2,3-dihydro-benzofuran-5-yl-ethanone, 1-2,3-dihydrobenzo b furan-5-yl ethan-1-one, 1-2,3-dihydrobenzofuran-5-yl ethanone, 5-acetyl-2,3-dihydro-1-benzofuran, 5-acetyl-2,3-dihydrobenzo b furan, 5-acetyl-2,3-dihydrobenzofuran
CC(=O)C1=CC=C2OCCC2=C1
162.19
162.19
90843-31-5
120°C to 121°C (15mmHg)
MFCD00068033
0.5g
MMVUJVASBDVNGJ-UHFFFAOYSA-N
1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
145220
97%

Chemical Identifiers

90843-31-5
162.19
MMVUJVASBDVNGJ-UHFFFAOYSA-N
145220
CC(=O)C1=CC=C2OCCC2=C1
C10H10O2
MFCD00068033
1-2,3-dihydro-1-benzofuran-5-yl ethan-1-one, 1-2,3-dihydro-1-benzofuran-5-yl ethanone, 1-2,3-dihydro-5-benzofuranyl ethanone, 1-2,3-dihydro-benzofuran-5-yl-ethanone, 1-2,3-dihydrobenzo b furan-5-yl ethan-1-one, 1-2,3-dihydrobenzofuran-5-yl ethanone, 5-acetyl-2,3-dihydro-1-benzofuran, 5-acetyl-2,3-dihydrobenzo b furan, 5-acetyl-2,3-dihydrobenzofuran
1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications
Promotions

Promotions