Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetamidopyridine-3-boronic acid pinacol ester, 98%

Chemical Identifiers

Chemical Name or Material 5-Acetamidopyridine-3-boronic acid pinacol ester
CAS 1201645-46-6
Formula Weight 262.12
MDL Number MFCD11878288
Beilstein 19927460
Synonym N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-pyridyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66479MD

 
alfa aesar™
H66479MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.12
Chemical Name or Material 5-Acetamidopyridine-3-boronic acid pinacol ester
Physical Form Solid
MDL Number MFCD11878288
CAS 1201645-46-6
Beilstein 19927460
Synonym N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-pyridyl]acetamide
Quantity 250mg
Assay 98%