Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetamido-2-nitrobenzoic acid, 98%

$66.45 - $251.64

Chemical Identifiers

Chemical Name or Material 5-Acetamido-2-nitrobenzoic acid
CAS 4368-83-6
Molecular Formula C9H8N2O5
Formula Weight 224.17
MDL Number MFCD00035889
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2056809

  SDS
alfa aesar™
B2056809
10g Each for $66.45

AAB2056818

  SDS
alfa aesar™
B2056818
50g Each for $251.64
Description & Specifications

Specifications

Formula Weight 224.17
TSCA TSCA
Health Hazard 1 Warning
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H8N2O5
CAS 4368-83-6
Quantity 10g
MDL Number MFCD00035889
Chemical Name or Material 5-Acetamido-2-nitrobenzoic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.