Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Acetamido-2-bromobenzoic acid hydrate, 95%

$341.72 - $341.72

Chemical Identifiers

Chemical Name or Material 5-Acetamido-2-bromobenzoic acid hydrate
CAS 22921-67-1
Molecular Formula C9H8BrNO3·xH2O
Formula Weight 258.07 (Anhydrous)
MDL Number MFCD00040902
Beilstein 3286005
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0962614

  SDS
alfa aesar™
L0962614
25g Each for $341.72
Description & Specifications

Specifications

Formula Weight 258.07 (Anhydrous)
Chemical Name or Material 5-Acetamido-2-bromobenzoic acid hydrate
Molecular Formula C9H8BrNO3·xH2O
MDL Number MFCD00040902
CAS 22921-67-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Beilstein 3286005
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 186° to 188°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.