Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,8-Dibromoisoquinoline, 98%

Chemical Identifiers

Chemical Name or Material 5,8-Dibromoisoquinoline
CAS 81045-39-8
Formula Weight 286.95
MDL Number MFCD03109888
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6303403

  SDS
alfa aesar™
H6303403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63034MD

  SDS
alfa aesar™
H63034MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.95
Chemical Name or Material 5,8-Dibromoisoquinoline
Physical Form Solid
Assay 98%
MDL Number MFCD03109888
CAS 81045-39-8
Quantity 1g