Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,7-Dodecadiyne-1,12-diol, 96%

$74.45 - $74.45

Chemical Identifiers

Chemical Name or Material 5,7-Dodecadiyne-1,12-diol
CAS 74602-32-7
Molecular Formula C12H18O2
Formula Weight 194.28
MDL Number MFCD00078343
Beilstein 1757614
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1148503

  SDS
alfa aesar™
L1148503
1g Each for $74.45
Description & Specifications

Specifications

Formula Weight 194.28
MDL Number MFCD00078343
CAS 74602-32-7
Chemical Name or Material 5,7-Dodecadiyne-1,12-diol
Assay 96%
Molecular Formula C12H18O2
Beilstein 1757614
Melting Point 49° to 51°C
Quantity 1g