Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,7-Dimethylisatin, 96%

$84.82 - $857.35

Chemical Identifiers

Chemical Name or Material 5,7-Dimethylisatin
CAS 39603-24-2
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00047219
Beilstein 143679
Synonym 5,7-Dimethyl-2,3-indolinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1133203

  SDS
alfa aesar™
L1133203
1g Each for $84.82

AAL1133206

  SDS
alfa aesar™
L1133206
5g Each for $244.09

AAL1133214

  SDS
alfa aesar™
L1133214
25g Each for $857.35
Description & Specifications

Specifications

Formula Weight 175.19
Chemical Name or Material 5,7-Dimethylisatin
MDL Number MFCD00047219
Synonym 5,7-Dimethyl-2,3-indolinedione
Molecular Formula C10H9NO2
Assay 96%
RTECS Number NL7930000
Melting Point 244° to 248°C
Beilstein 143679
Quantity 1g
CAS 39603-24-2

Applications
5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.