missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,7-Dimethylisatin, 96%

Manufacturer:  Alfa Aesar™ L1133203

 View more versions of this product

Catalog No. AAL1133203



Description

Description

Applications
5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Specifications

5,7-Dimethylisatin
1g
C10H9NO2
143679
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
5,7-dimethyl-1H-indole-2,3-dione
38296
96%
244°C to 248°C
39603-24-2
MFCD00047219
5,7-dimethylisatin, 5,7-dimethylindoline-2,3-dione, 5,7-dimethyl-indole-2,3-dione, 5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 5,7-dimethyl, indole-2,3-dione, 5,7-dimethyl, 1h-indole-2,3-dione, 5,7-dimethyl-9ci, acmc-20a4zq, isatin-based compound, 48, 1h-indole-2,3-dione,5,7-dimethyl
CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C
175.187
175.19

Chemical Identifiers

39603-24-2
175.187
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
38296
CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C
C10H9NO2
MFCD00047219
5,7-dimethylisatin, 5,7-dimethylindoline-2,3-dione, 5,7-dimethyl-indole-2,3-dione, 5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 5,7-dimethyl, indole-2,3-dione, 5,7-dimethyl, 1h-indole-2,3-dione, 5,7-dimethyl-9ci, acmc-20a4zq, isatin-based compound, 48, 1h-indole-2,3-dione,5,7-dimethyl
5,7-dimethyl-1H-indole-2,3-dione
Safety and Handling

Safety and Handling

RTECSNumber : NL7930000

Documents