missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine, 98%, Thermo Scientific™
Chemical Identifiers
CAS
1913-72-0
Molecular Formula
C9H9N3O
Molecular Weight (g/mol)
175.19
MDL Number
MFCD00138563
InChI Key
HWGAGILTAOQBPD-UHFFFAOYSA-N
Synonym
5,7-dimethyl-1h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3-hydropyridino 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h,4h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-4-hydroxypyrido 2,3-d pyrimidine, 5,7-dimethyl-4-oxo-3,4-dihydropyrido 2,3-d pyrimidine, 5,7-dimethylpyrido 2,3-d pyrimidin-4 3h-one, 5?7-dimethyl-4-hydroxypyrido 2?3-d pyrimidine, pyrido 2,3-d pyrimidin-4 3h-one,5,7-dimethyl
PubChem CID
5394613
IUPAC Name
5,7-dimethyl-1H-pyrido[2,3-d]pyrimidin-4-one
SMILES
CC1=CC(C)=C2C(=O)N=CNC2=N1
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
CAS
1913-72-0
Molecular Weight (g/mol)
175.19
InChI Key
HWGAGILTAOQBPD-UHFFFAOYSA-N
PubChem CID
5394613
SMILES
CC1=CC(C)=C2C(=O)N=CNC2=N1
Molecular Formula
C9H9N3O
MDL Number
MFCD00138563
Synonym
5,7-dimethyl-1h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3-hydropyridino 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h,4h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-4-hydroxypyrido 2,3-d pyrimidine, 5,7-dimethyl-4-oxo-3,4-dihydropyrido 2,3-d pyrimidine, 5,7-dimethylpyrido 2,3-d pyrimidin-4 3h-one, 5?7-dimethyl-4-hydroxypyrido 2?3-d pyrimidine, pyrido 2,3-d pyrimidin-4 3h-one,5,7-dimethyl
IUPAC Name
5,7-dimethyl-1H-pyrido[2,3-d]pyrimidin-4-one
Specifications
CAS
1913-72-0
CAS Max %
100.0
Elemental Analysis
97.5% min.
Infrared Spectrum
Authentic
MDL Number
MFCD00138563
Synonym
5,7-dimethyl-1h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3-hydropyridino 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h,4h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-4-hydroxypyrido 2,3-d pyrimidine, 5,7-dimethyl-4-oxo-3,4-dihydropyrido 2,3-d pyrimidine, 5,7-dimethylpyrido 2,3-d pyrimidin-4 3h-one, 5?7-dimethyl-4-hydroxypyrido 2?3-d pyrimidine, pyrido 2,3-d pyrimidin-4 3h-one,5,7-dimethyl
InChI Key
HWGAGILTAOQBPD-UHFFFAOYSA-N
IUPAC Name
5,7-dimethyl-1H-pyrido[2,3-d]pyrimidin-4-one
PubChem CID
5394613
Physical Form
Amorphous Powder
Chemical Name or Material
5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine
CAS Min %
97.5
Color
Beige
Assay Percent Range
98%
Molecular Formula
C9H9N3O
Packaging
Glass bottle
Quantity
1g
SMILES
CC1=CC(C)=C2C(=O)N=CNC2=N1
Molecular Weight (g/mol)
175.19
Formula Weight
175.19
Percent Purity
98%
Safety and Handling
Safety and Handling
GHS H Statement: Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin.
GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
Product Certifications