missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,7-Dihydroindolo[2,3-b]carbazole, 96%, Alfa Aesar™

$282.20 - $869.76

Chemical Identifiers

CAS 111296-90-3
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.308
MDL Number MFCD10687177
InChI Key UVHIWKCVGUMHNB-UHFFFAOYSA-N
Synonym 5,7-dihydroindolo 2,3-b carbazole, 5,7-dihydro-indolo 2,3-b carbazole, indolo 2,3-b carbazole,5,7-dihydro, 5h,7h-indolo 2,3-b carbazole, pubchem23468, acmc-209wje, 7h-indolo 2,3-b carbazole, 1,3-dihydroindolo 2,3-b carbazole, indolo 2,3-arbazole, 5,7-ihydro, indolo 2,3-b carbazole, 5,7-dihydro
PubChem CID 10332633
IUPAC Name 5,7-dihydroindolo[2,3-b]carbazole
SMILES C1=CC=C2C(=C1)C3=CC4=C(C=C3N2)NC5=CC=CC=C54
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6605603 View Documents Alfa Aesar
H6605603
1g Each for $869.76
 
AAH66056MD View Documents Alfa Aesar
H66056MD
250mg Each for $282.20
 
Specifications

Chemical Identifiers

111296-90-3
256.308
UVHIWKCVGUMHNB-UHFFFAOYSA-N
10332633
C1=CC=C2C(=C1)C3=CC4=C(C=C3N2)NC5=CC=CC=C54
C18H12N2
MFCD10687177
5,7-dihydroindolo 2,3-b carbazole, 5,7-dihydro-indolo 2,3-b carbazole, indolo 2,3-b carbazole,5,7-dihydro, 5h,7h-indolo 2,3-b carbazole, pubchem23468, acmc-209wje, 7h-indolo 2,3-b carbazole, 1,3-dihydroindolo 2,3-b carbazole, indolo 2,3-arbazole, 5,7-ihydro, indolo 2,3-b carbazole, 5,7-dihydro
5,7-dihydroindolo[2,3-b]carbazole

Specifications

1.40g/mL
348°C to 360°C
111296-90-3
MFCD10687177
UVHIWKCVGUMHNB-UHFFFAOYSA-N
5,7-dihydroindolo[2,3-b]carbazole
10332633
Solid
5,7-Dihydroindolo[2,3-b]carbazole
Brown
1g
C18H12N2
5,7-dihydroindolo 2,3-b carbazole, 5,7-dihydro-indolo 2,3-b carbazole, indolo 2,3-b carbazole,5,7-dihydro, 5h,7h-indolo 2,3-b carbazole, pubchem23468, acmc-209wje, 7h-indolo 2,3-b carbazole, 1,3-dihydroindolo 2,3-b carbazole, indolo 2,3-arbazole, 5,7-ihydro, indolo 2,3-b carbazole, 5,7-dihydro
C1=CC=C2C(=C1)C3=CC4=C(C=C3N2)NC5=CC=CC=C54
256.308
256.3
96%