missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,7-Dichlorokynurenic acid sodium salt, Thermo Scientific™
Potent NMDA receptor glycine site antagonist
$812.51 - $812.51
Chemical Identifiers
CAS | 1184986-70-6 |
---|---|
Molecular Formula | C10H4Cl2NNaO3 |
Molecular Weight (g/mol) | 280.036 |
MDL Number | MFCD00674925 |
InChI Key | VPRPMJHKWHCUFW-UHFFFAOYSA-M |
Synonym | 5,7-dichlorokynurenic acid sodium salt, sodium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, potassium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid sodium salt |
PubChem CID | 44134672 |
IUPAC Name | sodium;5,7-dichloro-4-oxo-1H-quinoline-2-carboxylate |
SMILES | C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)[O-])Cl.[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61567MF
|
Thermo Scientific Chemicals
J61567MF |
50 mg |
Each for $812.51
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications5,7-Dichlorokynurenic acid sodium salt is a potent NMDA receptor glycine site antagonist. It acts as a chemical and organic intermediate.
Solubility
Soluble in DMSO. Insoluble in water
Notes
May be stored at 4°C. For long-term storage, aliquot and store at 4°C. Do not freeze. For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer. Protect from light.
Chemical Identifiers
1184986-70-6 | |
280.036 | |
VPRPMJHKWHCUFW-UHFFFAOYSA-M | |
44134672 | |
C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)[O-])Cl.[Na+] |
C10H4Cl2NNaO3 | |
MFCD00674925 | |
5,7-dichlorokynurenic acid sodium salt, sodium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, potassium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid sodium salt | |
sodium;5,7-dichloro-4-oxo-1H-quinoline-2-carboxylate |
Specifications
1184986-70-6 | |
C10H4Cl2NNaO3 | |
50 mg | |
Soluble in DMSO; Insoluble in water | |
C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)[O-])Cl.[Na+] | |
280.036 | |
280.04 | |
5,7-Dichlorokynurenic acid sodium salt |
Yellow | |
MFCD00674925 | |
5,7-dichlorokynurenic acid sodium salt, sodium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, potassium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid sodium salt | |
VPRPMJHKWHCUFW-UHFFFAOYSA-M | |
sodium;5,7-dichloro-4-oxo-1H-quinoline-2-carboxylate | |
44134672 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : VB1984600
TSCA : No
Recommended Storage : Keep cold