missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,7-Dichloro-8-hydroxyquinoline 98.0+%, TCI America™

$98.50 - $261.11

Chemical Identifiers

CAS 773-76-2
Molecular Formula C9H5Cl2NO
Molecular Weight (g/mol) 214.045
MDL Number MFCD00006786
InChI Key WDFKMLRRRCGAKS-UHFFFAOYSA-N
Synonym 5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
PubChem CID 2722
ChEBI CHEBI:59477
IUPAC Name 5,7-dichloroquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D04125G
View Documents
Tci America
D04125G
5g
Each for $98.50
 
D041225G
View Documents
Tci America
D041225G
25g
Each for $261.11
 
Description

Description

Specifications

Chemical Identifiers

773-76-2
214.045
WDFKMLRRRCGAKS-UHFFFAOYSA-N
2722
5,7-dichloroquinolin-8-ol
C9H5Cl2NO
MFCD00006786
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
CHEBI:59477
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1

Specifications

White-Yellow
5g
C9H5Cl2NO
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1
214.045
CHEBI:59477
Crystal-Powder at 20°C
5,7-Dichloro-8-hydroxyquinoline
181°C
773-76-2
MFCD00006786
WDFKMLRRRCGAKS-UHFFFAOYSA-N
5,7-dichloroquinolin-8-ol
2722
214.05
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : VC5350000

TSCA : Yes

SDS