missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,7-Dichloro-8-hydroxyquinoline, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113431000
Specifications
5, 7-Dichloro-8-hydroxyquinoline | |
Glass bottle | |
178°C to 183°C | |
99% | |
98.5 | |
97.5% min. (GC) | |
MFCD00006786 | |
15, 2181 | |
Solubility in water: practically insoluble. Other solubilities: soluble in water at ph 1 or >9;,slightly soluble in ethanol,acetone,chloroform,,and dmso;,soluble in alkali,benzene,petroleum ether | |
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1 | |
214.05 | |
CHEBI:59477 | |
Fluffy Powder |
Authentic | |
Beige | |
100g | |
773-76-2 | |
100.0 | |
C9H5Cl2NO | |
21, 95 | |
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin | |
WDFKMLRRRCGAKS-UHFFFAOYSA-N | |
5,7-dichloroquinolin-8-ol | |
2722 | |
214.05 | |
98% |
Chemical Identifiers
773-76-2 | |
214.05 | |
WDFKMLRRRCGAKS-UHFFFAOYSA-N | |
2722 | |
5,7-dichloroquinolin-8-ol |
C9H5Cl2NO | |
MFCD00006786 | |
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin | |
CHEBI:59477 | |
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 212-258-3
RTECSNumber : VC5350000
TSCA : TSCA