missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,7-Dichloro-8-hydroxyquinoline, 98%, ACROS Organics™

$18.49 - $18.49

Chemical Identifiers

CAS 773-76-2
Molecular Formula C9H5Cl2NO
Molecular Weight (g/mol) 214.05
MDL Number MFCD00006786
InChI Key WDFKMLRRRCGAKS-UHFFFAOYSA-N
Synonym 5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
PubChem CID 2722
ChEBI CHEBI:59477
IUPAC Name 5,7-dichloroquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC113430050
SDS View Documents
Thermo Fisher Scientific
113430050
5g Glass bottle
Each for $18.49
 
AC113431000
SDS View Documents
Thermo Fisher Scientific
113431000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

773-76-2
214.05
WDFKMLRRRCGAKS-UHFFFAOYSA-N
2722
5,7-dichloroquinolin-8-ol
C9H5Cl2NO
MFCD00006786
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
CHEBI:59477
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1

Specifications

Authentic
Beige
5g
98.5
97.5% min. (GC)
MFCD00006786
15, 2181
Solubility in water: practically insoluble. Other solubilities: soluble in water at ph 1 or >9;,slightly soluble in ethanol,acetone,chloroform,,and dmso;,soluble in alkali,benzene,petroleum ether
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1
214.05
CHEBI:59477
Fluffy Powder
5, 7-Dichloro-8-hydroxyquinoline, 99%
Glass bottle
178°C to 183°C
773-76-2
100.0
C9H5Cl2NO
21, 95
5,7-dichloro-8-hydroxyquinoline, 5,7-dichloro-8-quinolinol, capitrol, chloroxine, chloroxyquinoline, chlorquinol, dichloroquinolinol, dichloroxin, dikhloroskin, quixalin
WDFKMLRRRCGAKS-UHFFFAOYSA-N
5,7-dichloroquinolin-8-ol
2722
214.05
98%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 212-258-3

RTECSNumber : VC5350000

TSCA : TSCA

SDS