Learn More
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2406603
Description
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4″-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).
Solubility
Soluble in water (partly).
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid monosodium salt hydrate | |
C20H15N4NaO7S2 | |
1 g | |
ferrozine mono-sodium salt hydrate, pdt disulfonate monosodium salt hydrate, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate, sodium 4-3-pyridin-2-yl-6-4-sulfophenyl-1,2,4-triazin-5-yl benzenesulfonate hydrate | |
MBOKSYFCYXIJRZ-UHFFFAOYSA-M | |
sodium;4-[3-pyridin-2-yl-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate | |
71311205 | |
97% |
1266615-85-3 | |
MFCD00150794 | |
5222087 | |
Soluble in water (partly). | |
C1=CC=NC(=C1)C2=NC(=C(N=N2)C3=CC=C(C=C3)S(=O)(=O)O)C4=CC=C(C=C4)S(=O)(=O)[O-].O.[Na+] | |
510.471 | |
492.47 (Anhydrous) |
Chemical Identifiers
1266615-85-3 | |
510.471 | |
MBOKSYFCYXIJRZ-UHFFFAOYSA-M | |
71311205 | |
C1=CC=NC(=C1)C2=NC(=C(N=N2)C3=CC=C(C=C3)S(=O)(=O)O)C4=CC=C(C=C4)S(=O)(=O)[O-].O.[Na+] |
C20H15N4NaO7S2 | |
MFCD00150794 | |
ferrozine mono-sodium salt hydrate, pdt disulfonate monosodium salt hydrate, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate, sodium 4-3-pyridin-2-yl-6-4-sulfophenyl-1,2,4-triazin-5-yl benzenesulfonate hydrate | |
sodium;4-[3-pyridin-2-yl-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 274-196-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only