Please login to your online account to display your discounted pricing

5,6-Diphenyl-1,2,4-triazin-3-amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
CAS 4511-99-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H12N4
Formula Weight 248.29
MDL Number MFCD00047461
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01743EA

 
fisher scientific company
SB01743EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SB01743DA

 
fisher scientific company
SB01743DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.29
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00047461
CAS 4511-99-3
Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
Molecular Formula C15H12N4