Please login to your online account to display your discounted pricing

5,6-Diphenyl-1,2,4-triazin-3-amine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB01743EA

 View more versions of this product

Catalog No. SB01743EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Amber Glass Bottle
Quantity 10g
Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
CAS 4511-99-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H12N4
Formula Weight 248.29g/mol
MDL Number MFCD00047461

Specifications

Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
CAS 4511-99-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H12N4
Formula Weight 248.29g/mol
MDL Number MFCD00047461