Please login to your online account to display your discounted pricing

5,6-Diphenyl-1,2,4-triazin-3-amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB01743DA

 View more versions of this product

Catalog No. SB01743DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 248.29
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C15H12N4
CAS 4511-99-3
MDL Number MFCD00047461
Packaging Amber Glass Bottle
Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
Assay Percent Range 97%

Specifications

Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
CAS 4511-99-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H12N4
Formula Weight 248.29
MDL Number MFCD00047461