missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6-Dimethylbenzimidazole, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115980250

 View more versions of this product

Catalog No. AC115980250



Description

Description

Specifications

Specifications

5, 6-Dimethylbenzimidazole
Glass bottle
203°C to 206°C
99+%
99.0
99+%
MFCD00005603
1h-benzimidazole, 5,6-dimethyl, 5,6-dimethyl-1h-benzo d imidazole, 5,6-dimethylbenzimidazole, 5.6-dimethylbenzimidazole, benzimidazole, 5,6-dimethyl, dimedazol, dimedazole, dimesol, dimethylbenzimidazole, dimezol
LJUQGASMPRMWIW-UHFFFAOYSA-N
5,6-dimethyl-1H-benzimidazole
675
146.19
99+%
Authentic
Beige
25g
582-60-5
100.0
C9H10N2
15, 3258
Solubility in water: soluble. Other solubilities: freely soluble in diluted acids,soluble in chloroform,ether and ethanol
CC1=CC2=C(C=C1C)N=CN2
146.19
CHEBI:15890
Powder

Chemical Identifiers

582-60-5
146.19
LJUQGASMPRMWIW-UHFFFAOYSA-N
675
5,6-dimethyl-1H-benzimidazole
C9H10N2
MFCD00005603
1h-benzimidazole, 5,6-dimethyl, 5,6-dimethyl-1h-benzo d imidazole, 5,6-dimethylbenzimidazole, 5.6-dimethylbenzimidazole, benzimidazole, 5,6-dimethyl, dimedazol, dimedazole, dimesol, dimethylbenzimidazole, dimezol
CHEBI:15890
CC1=CC2=C(C=C1C)N=CN2
Safety and Handling

Safety and Handling

EINECSNumber : 209-488-1

RTECSNumber : DD8280000

TSCA : TSCA

SDS Documents