Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethylbenzimidazole, 99+%

$45.09 - $132.07

Chemical Identifiers

Chemical Name or Material 5,6-Dimethylbenzimidazole
CAS 582-60-5
Molecular Formula C9H10N2
Formula Weight 146.19
MDL Number MFCD00005603
Beilstein 116595
Merck Index 14,3236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0746914

 
alfa aesar™
L0746914
25g Each for $45.09

AAL0746922

 
alfa aesar™
L0746922
100g Each for $132.07
Description & Specifications

Specifications

Formula Weight 146.19
Chemical Name or Material 5,6-Dimethylbenzimidazole
TSCA TSCA
Assay ≥99%
Molecular Formula C9H10N2
Melting Point 202° to 206°C
CAS 582-60-5
Merck Index 14,3236
RTECS Number DD8280000
MDL Number MFCD00005603
Beilstein 116595
Quantity 25g

Applications
5,6-dimethylbenzimidazole is one of the building blocks required for the synthesis of coenzyme B12. 5,6-Dimethylbenzimidazole is biosynthesized from flavin mononucleotide by the enzyme 5,6-dimethylbenzimidazole synthase

Solubility
Soluble in ethanol (50 mg/ml).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.