Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethylbenzimidazole-1-acetic acid

$83.47 - $83.47

Chemical Identifiers

Chemical Name or Material 5,6-Dimethylbenzimidazole-1-acetic acid
CAS 500872-62-8
Molecular Formula C11H12N2O2
Formula Weight 204.23g/mol
MDL Number MFCD08061049
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50893MD

 
alfa aesar™
H50893-MD
250mg Each for $83.47
Description & Specifications

Specifications

Quantity 250mg
Quantity 250mg
Chemical Name or Material 5,6-Dimethylbenzimidazole-1-acetic acid
CAS 500872-62-8
Molecular Formula C11H12N2O2
Formula Weight 204.23g/mol
MDL Number MFCD08061049
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.