Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethyl-2,1,3-benzothiadiazole, 97%

$134.41 - $527.36

Chemical Identifiers

Chemical Name or Material 5,6-Dimethyl-2,1,3-benzothiadiazole
CAS 1887-60-1
Molecular Formula C8H8N2S
Formula Weight 164.23
MDL Number MFCD00174277
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2075503

  SDS
alfa aesar™
B2075503
1g Each for $134.41

AAB2075506

  SDS
alfa aesar™
B2075506
5g Each for $527.36
Description & Specifications

Specifications

Formula Weight 164.23
Chemical Name or Material 5,6-Dimethyl-2,1,3-benzothiadiazole
Assay 97%
Molecular Formula C8H8N2S
Melting Point 83° to 85°C
MDL Number MFCD00174277
CAS 1887-60-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.