Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethyl-1H-benzotriazole monohydrate, 99%

$49.11 - $49.11

Chemical Identifiers

Chemical Name or Material 5,6-Dimethyl-1H-benzotriazole monohydrate
CAS 4184-79-6
Molecular Formula C8H9N3·H2O
Formula Weight 165.20 (147.18 Anhydrous)
MDL Number MFCD00149344
Beilstein 122933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2530406

  SDS
alfa aesar™
B2530406
5g Each for $49.11

AAB2530414

  SDS
alfa aesar™
B2530414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5,6-Dimethyl-1H-benzotriazole monohydrate
CAS 4184-79-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Molecular Formula C8H9N3·H2O
Melting Point 153° to 157°C
Formula Weight 165.20 (147.18 Anhydrous)
MDL Number MFCD00149344
Beilstein 122933
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.