missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,6-Dimethyl-1H-benzotriazole hydrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116050050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5, 6-Dimethyl-1H-benzotriazole hydrate | |
4184-79-6 | |
100.0 | |
99% | |
C8H9N3·xH2O | |
5g | |
VVWPOQUENPOMAK-UHFFFAOYSA-N | |
5,6-dimethyl-2H-benzotriazole;hydrate | |
16219214 | |
99% |
99% | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00149344 | |
5,6-dimethyl-1h-benzotriazole hydrate, 5,6-dimethyl-1,2,3-benzotriazole hydrate, 5,6-dimethyl-1h-1,2,3-benzotriazole hydrate, acmc-1aqbb, 1h-benzotriazole,5,6-dimethyl, 5,6-dimethylbenzotriazole, oxamethane, 5,6-dimethyl-2h-benzotriazole hydrate, 5,6-dimethyl-1h-benzotriazole monohydrate, 5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate | |
CC1=CC2=NNN=C2C=C1C.O | |
147.18 | |
147.18 |
Chemical Identifiers
4184-79-6 | |
147.18 | |
VVWPOQUENPOMAK-UHFFFAOYSA-N | |
16219214 | |
CC1=CC2=NNN=C2C=C1C.O |
C8H9N3·xH2O | |
MFCD00149344 | |
5,6-dimethyl-1h-benzotriazole hydrate, 5,6-dimethyl-1,2,3-benzotriazole hydrate, 5,6-dimethyl-1h-1,2,3-benzotriazole hydrate, acmc-1aqbb, 1h-benzotriazole,5,6-dimethyl, 5,6-dimethylbenzotriazole, oxamethane, 5,6-dimethyl-2h-benzotriazole hydrate, 5,6-dimethyl-1h-benzotriazole monohydrate, 5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate | |
5,6-dimethyl-2H-benzotriazole;hydrate |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 224-058-3
RUO â Research Use Only