missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6-Dimethyl-1H-benzotriazole Hydrate 99%, ACROS Organics™

Chemical Identifiers

CAS 4184-79-6
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H9N3·xH2O
Formula Weight 147.18
MDL Number MFCD00149344
Chemical Name or Material 5, 6-Dimethyl-1H-benzotriazole hydrate, 99%
Assay ≥98.5% (on dry substance)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC116050250 SDS Thermo Fisher Scientific
116050250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC116050050 SDS Thermo Fisher Scientific
116050050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4184-79-6
100.0
C8H9N3·xH2O
MFCD00149344
≥98.5% (on dry substance)
98.5
99%
147.18
5, 6-Dimethyl-1H-benzotriazole hydrate, 99%

Specifications

Authentic
153° to 156°C
4184-79-6
100.0
11% max. (K.F.)
99%
147.18
5, 6-Dimethyl-1H-benzotriazole hydrate, 99%
Yellow to brown
25g
98.5
Glass bottle
Crystalline Powder
C8H9N3·xH2O
MFCD00149344
≥98.5% (on dry substance)

Safety and Handling

EINECSNumber : 224-058-3