Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethyl-1,10-phenanthroline, 98%

$400.80 - $1,331.97

Chemical Identifiers

Chemical Name or Material 5,6-Dimethyl-1,10-phenanthroline
CAS 3002-81-1
Molecular Formula C14H12N2
Formula Weight 208.26
MDL Number MFCD00004983
Beilstein 160918
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2371703

  SDS
alfa aesar™
B2371703
1g Each for $400.80

AAB2371706

  SDS
alfa aesar™
B2371706
5g Each for $1,331.97

AAB23717MD

  SDS
alfa aesar™
B23717MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5,6-Dimethyl-1,10-phenanthroline
CAS 3002-81-1
TSCA TSCA
Assay 98%
Molecular Formula C14H12N2
Melting Point 266° to 269°C
Formula Weight 208.26
MDL Number MFCD00004983
Beilstein 160918
Quantity 1g