missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,6-Dimethoxyindole, 99%, ACROS Organics™
Chemical Identifiers
CAS | 14430-23-0 |
---|---|
Molecular Formula | C10H11NO2 |
Molecular Weight (g/mol) | 177.2 |
MDL Number | MFCD00005675 |
InChI Key | QODBZRNBPUPLEZ-UHFFFAOYSA-N |
Synonym | #, 1h-indole, 5,6-dimethoxy, 1h-indole,5,6-dimethoxy, 5,6-dimethoxyindol, 5,6-dimethoxyindole, acmc-1cbhq, pubchem7329, zlchem 178 |
PubChem CID | 84431 |
IUPAC Name | 5,6-dimethoxy-1H-indole |
SMILES | COC1=C(C=C2C(=C1)C=CN2)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397730010
|
Thermo Fisher Scientific
AC397730010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC397730050
|
Thermo Fisher Scientific
AC397730050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
14430-23-0 | |
177.2 | |
QODBZRNBPUPLEZ-UHFFFAOYSA-N | |
84431 | |
COC1=C(C=C2C(=C1)C=CN2)OC |
C10H11NO2 | |
MFCD00005675 | |
#, 1h-indole, 5,6-dimethoxy, 1h-indole,5,6-dimethoxy, 5,6-dimethoxyindol, 5,6-dimethoxyindole, acmc-1cbhq, pubchem7329, zlchem 178 | |
5,6-dimethoxy-1H-indole |
Specifications
Authentic | |
198.0°C (8.0 mmHg) | |
154.0°C | |
14430-23-0 | |
100.0 | |
C10H11NO2 | |
#, 1h-indole, 5,6-dimethoxy, 1h-indole,5,6-dimethoxy, 5,6-dimethoxyindol, 5,6-dimethoxyindole, acmc-1cbhq, pubchem7329, zlchem 178 | |
QODBZRNBPUPLEZ-UHFFFAOYSA-N | |
5,6-dimethoxy-1H-indole | |
84431 | |
Powder | |
5, 6-Dimethoxyindole |
Glass bottle | |
Beige | |
1g | |
98.5 | |
98.5% min. (HPLC) | |
MFCD00005675 | |
Solubility in water: insoluble | |
COC1=C(C=C2C(=C1)C=CN2)OC | |
177.2 | |
177.2 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 238-402-5