Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dimethoxy-2-phenylindole, 97%

$42.47 - $153.93

Chemical Identifiers

Chemical Name or Material 5,6-Dimethoxy-2-phenylindole
CAS 62663-26-7
Molecular Formula C16H15NO2
Formula Weight 253.3
MDL Number MFCD00798603
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2109102

  SDS
alfa aesar™
B2109102
0.5g Each for $153.93

AAB2109177

  SDS
alfa aesar™
B2109177
0.1g Each for $42.47
Description & Specifications

Specifications

Formula Weight 253.3
Chemical Name or Material 5,6-Dimethoxy-2-phenylindole
Assay 97%
Molecular Formula C16H15NO2
MDL Number MFCD00798603
CAS 62663-26-7
Quantity 0.5g