Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dihydro-6-methyluracil, 99%

$32.09 - $32.09

Chemical Identifiers

Chemical Name or Material 5,6-Dihydro-6-methyluracil
CAS 2434-49-3
Molecular Formula C5H8N2O2
Formula Weight 128.13
MDL Number MFCD00023160
Beilstein 118196
Synonym 6-Methyl-5,6-dihydrouracil
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0229203

  SDS
alfa aesar™
L0229203
1g Each for $32.09
Description & Specifications

Specifications

Formula Weight 128.13
MDL Number MFCD00023160
Chemical Name or Material 5,6-Dihydro-6-methyluracil
Molecular Formula C5H8N2O2
CAS 2434-49-3
Assay 99%
Synonym 6-Methyl-5,6-dihydrouracil
Beilstein 118196
Quantity 1g
Melting Point 218° to 220°C